(3-Furan-2-yl-5-thioxo-1,5-dihydro-[1,2,4]triazol-4-yl)-acetic acid
Lieferant: Apollo Scientific
OR309410-1G
APOSOR309410-1GEA
475
EUR
APOSOR309410-1G
(3-Furan-2-yl-5-thioxo-1,5-dihydro-[1,2,4]triazol-4-yl)-acetic acid
(3-Furan-2-yl-5-thioxo-1,5-dihydro-[1,2,4]triazol-4-yl)-acetic acid
Formel:
C₈H₇N₃O₃S Molecular Weight: 225,224 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08437536 CAS-Nummer: 110167-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...