(3-Thiophen-2-ylmethyl-5-thioxo-1,5-dihydro-[1,2,4]triazol-4-yl)-acetic acid

Lieferant: Apollo Scientific

OR309411-1G
APOSOR309411-1GEA 475 EUR
APOSOR309411-1G
(3-Thiophen-2-ylmethyl-5-thioxo-1,5-dihydro-[1,2,4]triazol-4-yl)-acetic acid
(3-Thiophen-2-ylmethyl-5-thioxo-1,5-dihydro-[1,2,4]triazol-4-yl)-acetic acid
Formel: C₉H₉N₃O₂S₂
Molecular Weight: 255,317 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08437537
CAS-Nummer: 892874-27-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR