2-(5-Methanesulphonyl-3-phenyl-[1,2,4]triazol-4-yl)-acetic acid

Lieferant: Apollo Scientific

OR309412-1G
APOSOR309412-1GEA 475 EUR
APOSOR309412-1G
2-(5-Methanesulphonyl-3-phenyl-[1,2,4]triazol-4-yl)-acetic acid
2-(5-Methanesulphonyl-3-phenyl-[1,2,4]triazol-4-yl)-acetic acid
Formel: C₁₁H₁₁N₃O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD10703489
CAS-Nummer: 892874-30-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR