5-Chloro-3-[(4-methoxyphenyl)methyl]-1,2,4-thiadiazole

Lieferant: Apollo Scientific

OR309413-1G
APOSOR309413-1GEA 467 EUR
APOSOR309413-1G
5-Chloro-3-[(4-methoxyphenyl)methyl]-1,2,4-thiadiazole
5-Chloro-3-[(4-methoxyphenyl)methyl]-1,2,4-thiadiazole
Formel: C₁₀H₉ClN₂OS
Lagertemperatur: Raumtemperatur
MDL: MFCD14707338
CAS-Nummer: 1029718-72-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR