5-Chloro-3-(methylthio)-1,2,4-thiadiazole

Lieferant: Apollo Scientific

OR309420-1G
APOSOR309420-1GEA 467 EUR
APOSOR309420-1G
5-Chloro-3-(methylthio)-1,2,4-thiadiazole
5-Chloro-3-(methylthio)-1,2,4-thiadiazole
Formel: C₃H₃ClN₂S₂
Molecular Weight: 166,652 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07779433
CAS-Nummer: 21735-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR