Boc-DL-β-Aib-OH

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-((tert-Butoxycarbonyl)amino)-2-methylpropanoic acid

OR309427-250MG
APOSOR309427-250MGEA 46.4 EUR
APOSOR309427-250MG
Boc-DL-β-Aib-OH
Boc-DL-beta-Aib-OH
Formel: C₉H₁₇NO₄
Molecular Weight: 203,24 g/mol
Schmelzpunkt: 88…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02094137
CAS-Nummer: 16948-10-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR