4-Morpholino-2-pyridinamin

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-4-(morpholin-4-yl)pyridine, 4-Morpholinopyridin-2-amine

OR30943-1G
APOSOR30943-1GEA 0 EUR
APOSOR30943-1G
4-Morpholino-2-pyridinamin
4-Morpholino-2-pyridinamin
Formel: C₉H₁₃N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD09991750
CAS-Nummer: 722549-98-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR