(1S,2S)-1-Amino-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid hydrochloride
Lieferant: Apollo Scientific
OR309434-100MG
APOSOR309434-100MGEA
507
EUR
APOSOR309434-100MG
(1S,2S)-1-Amino-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid hydrochloride
(1S,2S)-1-Amino-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid hydrochloride
Formel:
C₁₁H₁₃NO₂·ClH Molecular Weight: 227,687 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09787518 CAS-Nummer: 888323-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...