(1R,2R,5S,7R)-N-tert-Butoxycarbonyl-8,8-dimethyl-3-azatricyclo[5.1.1.0

Lieferant: Apollo Scientific

OR309436-100MG
APOSOR309436-100MGEA 417 EUR
APOSOR309436-100MG
(1R,2R,5S,7R)-N-tert-Butoxycarbonyl-8,8-dimethyl-3-azatricyclo[5.1.1.0
(1R,2R,5S,7R)-N-tert-Butoxycarbonyl-8,8-dimethyl-3-azatricyclo[5.1.1.0^2,5]nonan-4-one
Formel: C₁₅H₂₃NO₃
Molecular Weight: 265,348 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09881415
CAS-Nummer: 1027343-55-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR