Ethyl (1R,2R,3S,5R)-2-amino-6,6-dimethylbicyclo[3.1.1]heptan-3-carboxylate hydrochloride

Lieferant: Apollo Scientific

OR309437-100MG
APOSOR309437-100MGEA 417 EUR
APOSOR309437-100MG
Ethyl (1R,2R,3S,5R)-2-amino-6,6-dimethylbicyclo[3.1.1]heptan-3-carboxylate hydrochloride
Ethyl (1R,2R,3S,5R)-2-amino-6,6-dimethylbicyclo[3.1.1]heptan-3-carboxylate hydrochloride
Formel: C₁₂H₂₁NO₂·ClH
Molecular Weight: 247,762 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09881416
CAS-Nummer: 1027343-58-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR