tert-Butyl (1R,2R,3S,5R)-(2-methylaminocarbonyl-6,6-dimethylbicyclo[3.1.1]hept-3-yl)-carbamate

Lieferant: Apollo Scientific

OR309438-100MG
APOSOR309438-100MGEA 417 EUR
APOSOR309438-100MG
tert-Butyl (1R,2R,3S,5R)-(2-methylaminocarbonyl-6,6-dimethylbicyclo[3.1.1]hept-3-yl)-carbamate
tert-Butyl (1R,2R,3S,5R)-(2-methylaminocarbonyl-6,6-dimethylbicyclo[3.1.1]hept-3-yl)-carbamate
Formel: C₁₆H₂₈N₂O₃
Molecular Weight: 296,405 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09881417
CAS-Nummer: 1027343-56-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR