(1R,2R,3S,5R)-2-Amino-6,6-dimethylbicyclo[3.1.1]heptan-3-carboxylic acid hydrochloride

Lieferant: Apollo Scientific

OR309439-100MG
APOSOR309439-100MGEA 409 EUR
APOSOR309439-100MG
(1R,2R,3S,5R)-2-Amino-6,6-dimethylbicyclo[3.1.1]heptan-3-carboxylic acid hydrochloride
(1R,2R,3S,5R)-2-Amino-6,6-dimethylbicyclo[3.1.1]heptan-3-carboxylic acid hydrochloride
Formel: C₁₀H₁₇NO₂·ClH
Molecular Weight: 219,708 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09881418
CAS-Nummer: 1027343-50-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR