4-Pyrrolidin-1-ylpyridin-2-amine

Lieferant: Apollo Scientific
Achtung

OR30944-1G
APOSOR30944-1GEA 360 EUR
APOSOR30944-1G
4-Pyrrolidin-1-ylpyridin-2-amine
4-Pyrrolidin-1-ylpyridin-2-amine
Formel: C₉H₁₃N₃
Lagertemperatur: Kühlschrank
MDL: MFCD08062967
CAS-Nummer: 722550-01-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR