(1R,2R,3S,5R)-(2-tert-Butoxycarbonylamino)-6,6-dimethylbicyclo[3.1.1]heptan-3-carboxylic acid

Lieferant: Apollo Scientific

OR309440-100MG
APOSOR309440-100MGEA 417 EUR
APOSOR309440-100MG
(1R,2R,3S,5R)-(2-tert-Butoxycarbonylamino)-6,6-dimethylbicyclo[3.1.1]heptan-3-carboxylic acid
(1R,2R,3S,5R)-(2-tert-Butoxycarbonylamino)-6,6-dimethylbicyclo[3.1.1]heptan-3-carboxylic acid
Formel: C₁₅H₂₅NO₄
Molecular Weight: 283,363 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09881419
CAS-Nummer: 1027343-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR