(1R,2R,3R,5R)-Ethyl-2-amino-6,6-dimethylbicyclo[3.1.1]heptan-3-carboxylate hydrochloride
Lieferant: Apollo Scientific
OR309441-100MG
APOSOR309441-100MGEA
517
EUR
APOSOR309441-100MG
(1R,2R,3R,5R)-Ethyl-2-amino-6,6-dimethylbicyclo[3.1.1]heptan-3-carboxylate hydrochloride
(1R,2R,3R,5R)-Ethyl-2-amino-6,6-dimethylbicyclo[3.1.1]heptan-3-carboxylate hydrochloride
Formel:
C₁₂H₂₁NO₂·ClH Molecular Weight: 247,762 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09881420 CAS-Nummer: 1027343-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...