cis-2-(9-Fluorenylmethoxycarbonylamino)cycloheptanecarboxylic acid

Lieferant: Apollo Scientific

OR309444-250MG
APOSOR309444-250MGEA 343 EUR
APOSOR309444-250MG
cis-2-(9-Fluorenylmethoxycarbonylamino)cycloheptanecarboxylic acid
cis-2-(9-Fluorenylmethoxycarbonylamino)cycloheptanecarboxylic acid
Formel: C₂₃H₂₅NO₄
Molecular Weight: 379,449 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09881424
CAS-Nummer: 1212250-46-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR