2-Chloro-6,8-dimethylquinoline-3-carbonitrile
Lieferant: Apollo Scientific
OR309452-1G
APOSOR309452-1GEA
0
EUR
APOSOR309452-1G
2-Chloro-6,8-dimethylquinoline-3-carbonitrile
2-Chloro-6,8-dimethylquinoline-3-carbonitrile
Formel:
C₁₂H₉ClN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08898633 CAS-Nummer: 917750-50-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...