5,7-Dichloro-2-methylquinoline-3-carboxylic acid

Lieferant: Apollo Scientific

OR309456-1G
APOSOR309456-1GEA 335 EUR
APOSOR309456-1G
5,7-Dichloro-2-methylquinoline-3-carboxylic acid
5,7-Dichloro-2-methylquinoline-3-carboxylic acid
Formel: C₁₁H₇Cl₂NO₂
Molecular Weight: 256,085 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787759
CAS-Nummer: 948293-69-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR