Ethyl-6-chlor-4-hydroxychinolin-3-carboxylat

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-4-hydroxyquinoline-3-carboxylic acid ethyl ester, Ethyl 6-chloro-4-hydroxyquinoline-3-carboxylate

OR309457-5G
APOSOR309457-5GEA 53.8 EUR
APOSOR309457-5G
Ethyl-6-chlor-4-hydroxychinolin-3-carboxylat
Ethyl-6-chlor-4-hydroxychinolin-3-carboxylat
Formel: C₁₂H₁₀ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00173342
CAS-Nummer: 70271-77-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR