5-Chlor-2,4,8-trimethylchinolin

Lieferant: Apollo Scientific

OR309459-1G
APOSOR309459-1GEA 293 EUR
APOSOR309459-1G
5-Chlor-2,4,8-trimethylchinolin
5-Chloro-2,4,8-trimethylquinoline
Formel: C₁₂H₁₂ClN
Lagertemperatur: Raumtemperatur
CAS-Nummer: 105908-43-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR