4-Bromo-7-methoxy-2-methylquinoline

Lieferant: Apollo Scientific

OR309461-1G
APOSOR309461-1GEA 383 EUR
APOSOR309461-1G
4-Bromo-7-methoxy-2-methylquinoline
4-Bromo-7-methoxy-2-methylquinoline
Formel: C₁₁H₁₀BrNO
Molecular Weight: 252,107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11505130
CAS-Nummer: 651042-71-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR