4-Chloro-8-ethyl-2-methylquinoline
Lieferant: Apollo Scientific
OR309464-1G
APOSOR309464-1GEA
301
EUR
APOSOR309464-1G
4-Chloro-8-ethyl-2-methylquinoline
4-Chloro-8-ethyl-2-methylquinoline
Formel:
C₁₂H₁₂ClN Molecular Weight: 205,683 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07644527 CAS-Nummer: 63136-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...