3-Bromo-4,6-dichloroquinoline

Lieferant: Apollo Scientific
Achtung

OR309467-1G
APOSOR309467-1GEA 359 EUR
APOSOR309467-1G
3-Bromo-4,6-dichloroquinoline
3-Bromo-4,6-dichloroquinoline
Formel: C₉H₄BrCl₂N
Molecular Weight: 276,945 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13192964
CAS-Nummer: 1204810-05-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR