3-Chloro-2-iodopyridine

Lieferant: Apollo Scientific
Achtung

OR30950-1G
APOSOR30950-1GEA 21.9 EUR
APOSOR30950-1G
3-Chloro-2-iodopyridine
3-Chloro-2-iodopyridine
Formel: C5H3ClIN
Molecular Weight: 239,4400 g/mol
Schmelzpunkt: 82…84 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09258810
CAS-Nummer: 77332-89-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR