4-(2-Chlorophenyl)-1,3-thiazol-2-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(2-Chlorophenyl)thiazol-2-amine

OR30955-1G
APOSOR30955-1GEA 21.9 EUR
APOSOR30955-1G
4-(2-Chlorophenyl)-1,3-thiazol-2-amine
4-(2-Chlorophenyl)-1,3-thiazol-2-amine
Formel: C₉H₇ClN₂S
Molecular Weight: 210,69 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01315352
CAS-Nummer: 21344-90-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR