tert-Butyl-4-[2-(4-amino-1H-pyrazol-1-yl)ethyl]piperazine-1-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-[2-(4-Amino-1H-pyrazol-1-yl)ethyl]piperazine, N1-BOC protected
OR30958-1G
APOSOR30958-1GEA
0
EUR
APOSOR30958-1G
tert-Butyl-4-[2-(4-amino-1H-pyrazol-1-yl)ethyl]piperazine-1-carboxylate
tert-Butyl-4-[2-(4-amino-1H-pyrazol-1-yl)ethyl]piperazine-1-carboxylate
Formel:
C₁₄H₂₅N₅O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11505005 CAS-Nummer: 1171901-11-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...