2-Amino-6-ethylpyrimidin-4-ol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-4-ethyl-6-hydroxypyrimidine
OR30961-250MG
APOSOR30961-250MGEA
42.2
EUR
APOSOR30961-250MG
2-Amino-6-ethylpyrimidin-4-ol
2-Amino-6-ethylpyrimidin-4-ol
Formel:
C₆H₉N₃O Molecular Weight: 139,16 g/mol Schmelzpunkt: 249…251 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10698994 CAS-Nummer: 5734-66-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...