2-Butyl-1,3-benzoxazol-6-amin
Lieferant: Apollo Scientific
OR30964-1G
APOSOR30964-1GEA
0
EUR
APOSOR30964-1G
2-Butyl-1,3-benzoxazol-6-amin
2-Butyl-1,3-benzoxazol-6-amin
Formel:
C₁₁H₁₄N₂O Lagertemperatur: Raumtemperatur |
CAS-Nummer:
875850-00-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...