2-Ethyl-1,3-benzoxazol-5-amin

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-2-ethyl-1,3-benzoxazole

OR30966-1G
APOSOR30966-1GEA 104 EUR
APOSOR30966-1G
2-Ethyl-1,3-benzoxazol-5-amin
2-Ethyl-1,3-benzoxazol-5-amin
Formel: C₉H₁₀N₂O
Lagertemperatur: Raumtemperatur
CAS-Nummer: 204771-75-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR