3-Amino-1-benzylpiperidine

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Benzylpiperidin-3-amine, 1-Benzylpiperidin-3-ylamine

OR30968-1G
APOSOR30968-1GEA 21.9 EUR
APOSOR30968-1G
3-Amino-1-benzylpiperidine
3-Amino-1-benzylpiperidine
Formel: C₁₂H₁₈N₂
Molecular Weight: 190,29 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04038444
CAS-Nummer: 60407-35-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR