6-Bromchinaldin

Lieferant: Apollo Scientific
Gefahr

OR3097-5G
APOSOR3097-5GEA 21.9 EUR
APOSOR3097-5G
6-Bromchinaldin
6-Bromchinaldin
Formel: C₁₀H₈BrN
Molecular Weight: 222,08 g/mol
Schmelzpunkt: 100…105 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00079724
CAS-Nummer: 877-42-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR