2-Methylimidazo[1,2-a]pyridin-3-amine 95%

Lieferant: Apollo Scientific
Achtung

OR30979-250MG
APOSOR30979-250MGEA 74.1 EUR
APOSOR30979-250MG
2-Methylimidazo[1,2-a]pyridin-3-amine 95%
2-Methylimidazo[1,2-a]pyridin-3-amine
Formel: C₈H₉N₃
Schmelzpunkt: 134…136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09800571
CAS-Nummer: 28036-31-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR