3-Amino-4-chlor-1-methyl-1H-indazol
Lieferant: Apollo Scientific
|
Achtung
|
OR30983-1G
APOSOR30983-1GEA
109
EUR
APOSOR30983-1G
3-Amino-4-chlor-1-methyl-1H-indazol
3-Amino-4-chloro-1-methyl-1H-indazole
Formel:
C₈H₈ClN₃ Schmelzpunkt: 122…124 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09864571 CAS-Nummer: 959240-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...