3-Amino-1H-indazol
Lieferant: Apollo Scientific
|
Achtung
|
OR30984-1G
OR30984-5G
OR30984-25G
APOSOR30984-1GEA
21.9
EUR
APOSOR30984-1G
APOSOR30984-5G
APOSOR30984-25G
3-Amino-1H-indazol
3-Amino-1H-indazole
Formel:
C₇H₇N₃ Schmelzpunkt: 156…158 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00182045 CAS-Nummer: 874-05-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...