2-Chloro-4-methylbenzaldehyde

Lieferant: Apollo Scientific
Achtung

OR30989-1G
APOSOR30989-1GEA 0 EUR
APOSOR30989-1G
2-Chloro-4-methylbenzaldehyde
2-Chloro-4-methylbenzaldehyde
Formel: C₈H₇ClO
Lagertemperatur: Raumtemperatur
MDL: MFCD10703515
CAS-Nummer: 50817-80-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR