5-(3-Oxo-3-phenyl-1-propenyl)-2-furaldehyde

Lieferant: Apollo Scientific

OR30990-1G
APOSOR30990-1GEA 0 EUR
APOSOR30990-1G
5-(3-Oxo-3-phenyl-1-propenyl)-2-furaldehyde
5-(3-Oxo-3-phenyl-1-propenyl)-2-furaldehyde
Formel: C₁₄H₁₀O₃
Molecular Weight: 226,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01315261
CAS-Nummer: 6575-75-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR