3-Chinolincarbaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
OR30994-50MG
OR30994-100MG
APOSOR30994-50MGEA
21.9
EUR
APOSOR30994-50MG
APOSOR30994-100MG
3-Chinolincarbaldehyd
3-Chinolincarbaldehyd
Formel:
C₁₀H₇NO Schmelzpunkt: 68…71 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006768 CAS-Nummer: 13669-42-6 EINECS: 237-147-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...