3-Chinolincarbaldehyd

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chinolincarboxaldehyd

OR30994-50MG OR30994-100MG
APOSOR30994-50MGEA 21.9 EUR
APOSOR30994-50MG APOSOR30994-100MG
3-Chinolincarbaldehyd
3-Chinolincarbaldehyd
Formel: C₁₀H₇NO
Molecular Weight: 157.17 g/mol
Schmelzpunkt: 68…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006768
CAS-Nummer: 13669-42-6
EINECS: 237-147-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR