3-Iodo-8-nitroimidazo[1,2-a]pyridine

Lieferant: Apollo Scientific

OR310007-1G OR310007-500MG
APOSOR310007-1GEA 336 EUR
APOSOR310007-1G APOSOR310007-500MG
3-Iodo-8-nitroimidazo[1,2-a]pyridine
3-Iodo-8-nitroimidazo[1,2-a]pyridine
Formel: C₇H₄IN₃O₂
Schmelzpunkt: 222…224 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD22126118
CAS-Nummer: 1373350-43-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR