Pyrido[3,4-d]pyrimidin-4(3H)-one

Lieferant: Apollo Scientific
Achtung

OR310008-1G
APOSOR310008-1GEA 81.2 EUR
APOSOR310008-1G
Pyrido[3,4-d]pyrimidin-4(3H)-one
Pyrido[3,4-d]pyrimidin-4(3H)-one
Formel: C7H5N3O
Molecular Weight: 147.1300 g/mol
Schmelzpunkt: 320 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD09999183
CAS-Nummer: 19178-25-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR