Ethyl-3-methylquinoxaline-2-carboxylate

Lieferant: Apollo Scientific

OR310043-1G
APOSOR310043-1GEA 0 EUR
APOSOR310043-1G
Ethyl-3-methylquinoxaline-2-carboxylate
Ethyl-3-methylquinoxaline-2-carboxylate
Formel: C₁₂H₁₂N₂O₂
Molecular Weight: 216.24 g/mol
Schmelzpunkt: 70…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02182648
CAS-Nummer: 3885-38-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR