(2S)-2-[(3aR,4R,6R,7aS)-3a,5,5-Trimethylhexahydro-4,6-methano-1,2-benzoxaborol-2(3H)-yl]pyrrolidine hydrochloride
Lieferant: Apollo Scientific
OR310083-500MG
APOSOR310083-500MGEA
1010
EUR
APOSOR310083-500MG
(2S)-2-[(3aR,4R,6R,7aS)-3a,5,5-Trimethylhexahydro-4,6-methano-1,2-benzoxaborol-2(3H)-yl]pyrrolidine hydrochloride
(2S)-2-[(3aR,4R,6R,7aS)-3a,5,5-Trimethylhexahydro-4,6-methano-1,2-benzoxaborol-2(3H)-yl]pyrrolidine hydrochloride
Formel:
C₁₅H₂₆BNO·ClH Molecular Weight: 283,645 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21602036 CAS-Nummer: 149716-72-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...