6-Ethoxyspiro[benzo[e][1,3]oxazine-2,4'-piperidin]-4(3H)-one hydrochloride

Lieferant: Apollo Scientific
Achtung

OR31010-250MG
APOSOR31010-250MGEA 464 EUR
APOSOR31010-250MG
6-Ethoxyspiro[benzo[e][1,3]oxazine-2,4'-piperidin]-4(3H)-one hydrochloride
6-Ethoxyspiro[benzo[e][1,3]oxazine-2,4'-piperidin]-4(3H)-one hydrochloride
Formel: C₁₄H₁₈N₂O₃·ClH
Molecular Weight: 298,765 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29042693
CAS-Nummer: 1823188-30-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR