N-(5-Iodo-3-methyl-pyridin-2-yl)-2,2-dimethylpropionamide
Lieferant: Apollo Scientific
Synonyme:
5-Iodo-3-methyl-2-(pivaloylamino)pyridine
OR310100-1G
APOSOR310100-1GEA
256
EUR
APOSOR310100-1G
N-(5-Iodo-3-methyl-pyridin-2-yl)-2,2-dimethylpropionamide
N-(5-Iodo-3-methyl-pyridin-2-yl)-2,2-dimethylpropionamide
Formel:
C₁₁H₁₅IN₂O Molecular Weight: 318.16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04218302 CAS-Nummer: 677327-29-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...