2-Bromo-5-(4-nitrophenoxy)-1,3,4-thiadiazole

Lieferant: Apollo Scientific

OR310136-1G OR310136-500MG
APOSOR310136-1GEA 671 EUR
APOSOR310136-1G APOSOR310136-500MG
2-Bromo-5-(4-nitrophenoxy)-1,3,4-thiadiazole
2-Bromo-5-(4-nitrophenoxy)-1,3,4-thiadiazole
Formel: C₈H₄BrN₃O₃S
Molecular Weight: 302.105 g/mol
Schmelzpunkt: 118…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD20368360
CAS-Nummer: 1343454-17-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR