2,2'-Dichlor-4,4'-bipyridyl
Lieferant: Apollo Scientific
Synonyme:
2,2'-Dichloro-4,4'-bipyridine
OR310158-100MG
APOSOR310158-100MGEA
0
EUR
APOSOR310158-100MG
2,2'-Dichlor-4,4'-bipyridyl
2,2'-Dichlor-4,4'-bipyridyl
Formel:
C₁₀H₆Cl₂N₂ Molecular Weight: 225,08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01106345 CAS-Nummer: 53344-74-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...