2-(4-Chlorophenyl)-2-oxoacetaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Chlorophenylglyoxal
OR31016-250MG
APOSOR31016-250MGEA
157
EUR
APOSOR31016-250MG
2-(4-Chlorophenyl)-2-oxoacetaldehyde
2-(4-Chlorophenyl)-2-oxoacetaldehyde
Formel:
C₈H₅ClO₂ Molecular Weight: 168,58 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03411516 CAS-Nummer: 4996-21-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...