3-Iodo-5-methylpyridin-2-ol

Lieferant: Apollo Scientific

Synonyme: 2-Hydroxy-3-iodo-5-methylpyridine

OR310163-1G
APOSOR310163-1GEA 63.8 EUR
APOSOR310163-1G
3-Iodo-5-methylpyridin-2-ol
3-Iodo-5-methylpyridin-2-ol
Formel: C₆H₆INO
Molecular Weight: 235.03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09037469
CAS-Nummer: 313678-93-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR