3-(1,3,4-Thiadiazol-2-yloxy)aniline

Lieferant: Apollo Scientific

OR310233-500MG
APOSOR310233-500MGEA 661 EUR
APOSOR310233-500MG
3-(1,3,4-Thiadiazol-2-yloxy)aniline
3-(1,3,4-Thiadiazol-2-yloxy)aniline
Formel: C₈H₇N₃OS
Molecular Weight: 193,226 g/mol
Schmelzpunkt: 123…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD19593858
CAS-Nummer: 1342452-85-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR