3-Chloro-5-(ethoxycarbonyl)pyridine-2-carboxylic acid

Lieferant: Apollo Scientific

OR310235-500MG
APOSOR310235-500MGEA 782 EUR
APOSOR310235-500MG
3-Chloro-5-(ethoxycarbonyl)pyridine-2-carboxylic acid
3-Chloro-5-(ethoxycarbonyl)pyridine-2-carboxylic acid
Formel: C₉H₈ClNO₄
Schmelzpunkt: 125…127 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD22373655
CAS-Nummer: 1200497-77-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR